Assay Details
Target Gene Details
Entrez Gene ID: | 20817 |
Gene Name: | serine/arginine-rich protein specific kinase 2 |
Gene Aliases: |
AW226533, AW492537, AW547358, Wbp6, mSRPK2 |
Location: |
Chr.5:23503356-23684641 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srpk2 | NM_009274.2 | 15 | 2799 | NP_033300.2 |
| AK028528.1 | 1 | 677 | BAC25993.1 | |
| AK032117.1 | 1 | 378 | ||
| AK033912.1 | 1 | 401 | ||
| AK035962.1 | 6 | 1210 | BAC29260.1 | |
| AK041426.1 | 5 | 1134 | BAC30942.1 | |
| AK082492.1 | 1 | 407 | ||
| AK136861.1 | 15 | 2765 | ||
| AK138155.1 | 1 | 308 | ||
| AK149055.1 | 4 | 940 | BAE28726.1 | |
| BC020178.1 | 15 | 2696 | AAH20178.1 | |
| BC062941.1 | 15 | 2718 | AAH62941.1 |
More Information
Set Membership: |
|

Genomic Map