Assay Details
Target Gene Details
Entrez Gene ID: | 94215 |
Gene Name: | UDP glucuronosyltransferase 2 family, polypeptide A1 |
Gene Aliases: |
- |
Location: |
Chr.5:87459490-87490871 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt2a1 | NM_053184.2 | 7 | 1776 | NP_444414.2 |
| AK140757.1 | 7 | 1777 | BAE24468.1 | |
| BC048926.1 | 6 | 1764 | AAH48926.1 |
Target Gene Details
Entrez Gene ID: | 552899 |
Gene Name: | UDP glucuronosyltransferase 2 family, polypeptide A2 |
Gene Aliases: |
- |
Location: |
Chr.5:87459493-87482258 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt2a2 | NM_001024148.1 | 7 | 1745 | NP_001019319.1 |
| BC058786.1 | 7 | 1745 | AAH58786.1 |

Genomic Map