Assay Details
Target Gene Details
Entrez Gene ID: | 77697 |
Gene Name: | methylmalonic aciduria (cobalamin deficiency) type B homolog (human) |
Gene Aliases: |
9130222L19Rik, ATR |
Location: |
Chr.5:114431034-114444073 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmab | NM_029956.3 | 9 | 2212 | NP_084232.1 |
| AK142286.1 | 8 | 1069 | BAE25010.1 | |
| BC057558.1 | 9 | 2212 | AAH57558.1 |
More Information
Set Membership: |
|

Genomic Map