Assay Details
Target Gene Details
Entrez Gene ID: | 319552 |
Gene Name: | RIKEN cDNA B230216G23 gene |
Gene Aliases: |
Npq, Spx |
Location: |
Chr.6:142410186-142419740 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B230216G23Rik | NM_001242345.1 | 6 | 1127 | NP_001229274.1 |
| AK043509.1 | 4 | 1040 | ||
| AK045625.1 | 3 | 1221 | ||
| BG092156.1 | 1 | 410 |

Genomic Map