Assay Details
Target Gene Details
Entrez Gene ID: | 106840 |
Gene Name: | unc-119 homolog B (C. elegans) |
Gene Aliases: |
AA407659, AI414892 |
Location: |
Chr.5:115122566-115134975 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc119b | NM_175352.4 | 5 | 1683 | NP_780561.1 |
| AK082614.1 | 5 | 1685 | BAC38549.1 | |
| BC008617.1 | 2 | 1153 | AAH08617.1 | |
| BC112388.1 | 5 | 1655 |
More Information
Set Membership: |
|

Genomic Map