Assay Details
Target Gene Details
Entrez Gene ID: | 66116 |
Gene Name: | camello-like 1 |
Gene Aliases: |
1110002I11Rik, AI266962 |
Location: |
Chr.6:85910154-85915677 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cml1 | NM_023160.2 | 2 | 874 | NP_075649.2 |
| AF163314.1 | 2 | 872 | AAF80483.1 | |
| AK002975.1 | 1 | 228 | BAB22489.1 | |
| AK003301.1 | 2 | 824 | BAB22702.1 | |
| AW045972.1 | 1 | 154 | ||
| BC010796.1 | 2 | 828 | AAH10796.1 |
More Information
Set Membership: |
|

Genomic Map