Assay Details
Target Gene Details
Entrez Gene ID: | 75379 |
Gene Name: | RIKEN cDNA 4930599N23 gene |
Gene Aliases: |
- |
Location: |
Chr.6:39118473-39148312 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930599N23Rik | NR_045813.1 | |||
| AK016417.1 | BAC25481.1 |
Target Gene Details
Entrez Gene ID: | 338523 |
Gene Name: | lysine (K)-specific demethylase 7A |
Gene Aliases: |
A630082K20Rik, BB041802, ENSMUSG00000073143, Jhdm1d, mKIAA1718 |
Location: |
Chr.6:39136620-39206773 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm7a | NM_001033430.4 | 20 | 3986 | NP_001028602.2 |
| AK035529.1 | 11 | 3000 | ||
| AK129429.1 | 9 | 2328 | BAC98239.1 | |
| AK136169.1 | 8 | 2125 | ||
| AK136238.1 | 20 | 3973 | BAE22890.1 | |
| BC145848.1 | 20 | 3876 |
More Information
Set Membership: |
|

Genomic Map