Assay Details
Target Gene Details
Entrez Gene ID: | 330277 |
Gene Name: | family with sequence similarity 71, member F1 |
Gene Aliases: |
GARI-L1, Gm764, NYD-SP18 |
Location: |
Chr.6:29313482-29336022 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam71f1 | NM_001289663.1 | 1 | 154 | NP_001276592.1 |
| NM_001289664.1 | 1 | 154 | NP_001276593.1 | |
| NM_001289665.1 | 1 | 154 | NP_001276594.1 | |
| NM_207258.3 | 1 | 154 | NP_997141.2 | |
| AB473725.1 | 1 | 95 | ||
| AK133908.1 | 1 | 155 | BAE21920.1 | |
| BC048651.1 | 1 | 155 | AAH48651.1 | |
| BC147513.1 | 1 | 137 | ||
| BC172074.1 | 1 | 137 | ||
| BC172086.1 | 1 | 137 |

Genomic Map