Assay Details
Target Gene Details
Entrez Gene ID: | 109880 |
Gene Name: | Braf transforming gene |
Gene Aliases: |
9930012E13Rik, AA120551, AA387315, AA473386, B-raf, Braf-2, Braf2, C230098H17, C87398, D6Ertd631e |
Location: |
Chr.6:39603231-39725656 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Braf | NM_139294.5 | 22 | 7350 | NP_647455.3 |
| AK049095.1 | 1 | 2076 | ||
| AK076310.1 | 1 | 585 | ||
| AK088830.1 | 1 | 2077 |
More Information
Set Membership: |
|

Genomic Map