Assay Details
Target Gene Details
Entrez Gene ID: | 654804 |
Gene Name: | RIKEN cDNA 4732471J01 gene |
Gene Aliases: |
- |
Location: |
Chr.7:25376819-25566417 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4732471J01Rik | NR_073027.1 | |||
| AK043644.1 | ||||
| AK076372.1 | ||||
| BB613302.1 |
Target Gene Details
Entrez Gene ID: | 16890 |
Gene Name: | lipase, hormone sensitive |
Gene Aliases: |
4933403G17Rik, HSL |
Location: |
Chr.7:25379527-25398704 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lipe | NM_001039507.2 | 2 | 497 | NP_001034596.1 |
| NM_010719.5 | 2 | 933 | NP_034849.2 | |
| AK019846.1 | 2 | 1334 | BAB31878.1 | |
| AK029984.1 | 2 | 936 | BAC26716.1 | |
| AK169858.1 | 2 | 492 | BAE41414.1 | |
| BC021642.1 | 2 | 493 | AAH21642.1 | |
| U08188.1 | 1 | 1017 | AAC52163.1 |
More Information
Set Membership: |
|

Genomic Map