Assay Details
Target Gene Details
Entrez Gene ID: | 56786 |
Gene Name: | TMEM9 domain family, member B |
Gene Aliases: |
2310004K06Rik, AW539847, D7H11orf15, ICRFP703B1614Q5.3 |
Location: |
Chr.7:109735834-109752870 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem9b | NM_020050.1 | 5 | 1499 | NP_064434.1 |
| AK009147.1 | BAB26105.1 | |||
| AK049882.1 | 5 | 1586 | ||
| AK077253.1 | 5 | 1455 | BAC36710.1 | |
| AK147127.1 | 5 | 1515 | BAE27698.1 | |
| AK194605.1 | ||||
| AK212155.1 | ||||
| AK216003.1 | ||||
| BC002208.1 | 5 | 1483 | AAH02208.1 |

Genomic Map