Assay Details
Target Gene Details
Entrez Gene ID: | 330483 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 16 |
Gene Aliases: |
Gm769 |
Location: |
Chr.7:19852097-19871449 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam16 | NM_001033419.2 | 3 | 670 | NP_001028591.2 |
| BC141087.1 | 3 | 581 |
Target Gene Details
Entrez Gene ID: | 100502736 |
Gene Name: | predicted gene, 19345 |
Gene Aliases: |
- |
Location: |
Chr.7:19852003-19858355 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm19345 | NM_001270489.1 | 4 | 604 | NP_001257418.1 |
| AK043814.1 | 4 | 605 | ||
| AW456762.1 | 1 | 404 | ||
| BI133264.1 |

Genomic Map