Assay Details
Target Gene Details
Entrez Gene ID: | 14282 |
Gene Name: | FBJ osteosarcoma oncogene B |
Gene Aliases: |
- |
Location: |
Chr.7:19302696-19310226 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fosb | NM_008036.2 | 1 | 253 | NP_032062.1 |
| AK154373.1 | 1 | 254 | ||
| BY337108.1 | 1 | 254 |
More Information
Set Membership: |
|

Genomic Map