Assay Details
Target Gene Details
Entrez Gene ID: | 14282 |
Gene Name: | FBJ osteosarcoma oncogene B |
Gene Aliases: |
- |
Location: |
Chr.7:19302696-19310226 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fosb | NM_008036.2 | 2 | 1012 | NP_032062.1 |
| BC132064.1 | 2 | 567 | ||
| BC137994.1 | 2 | 567 | ||
| CK795975.1 | 1 | 113 |
More Information
Set Membership: |
|

Genomic Map