Assay Details
Target Gene Details
Entrez Gene ID: | 65116 |
Gene Name: | proline-rich Gla (G-carboxyglutamic acid) polypeptide 2 |
Gene Aliases: |
1600014G02Rik, 2510041M06Rik, N4WBP1 |
Location: |
Chr.7:45053584-45062526 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrg2 | NM_022999.1 | 7 | 814 | NP_075375.1 |
| AF220205.1 | 3 | 343 | AAG44244.1 | |
| AK005453.1 | 8 | 857 | ||
| AK011086.2 | 1 | 81 | ||
| BC025098.1 | 7 | 814 | AAH25098.1 |
More Information
Set Membership: |
|

Genomic Map