Assay Details
Target Gene Details
Entrez Gene ID: | 75430 |
Gene Name: | anaphase prompoting complex C subunit 15 |
Gene Aliases: |
3200002M19Rik, 6330414C15Rik, APC15 |
Location: |
Chr.7:101881300-101901849 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc15 | AK018169.1 | BAB31105.1 | ||
| AK031167.1 | 1 | 394 | ||
| AK135201.1 | 6 | 1603 | BAE22458.1 |
Target Gene Details
Entrez Gene ID: | 791260 |
Gene Name: | transmembrane O-methyltransferase |
Gene Aliases: |
Comt2, F930017I19Rik |
Location: |
Chr.7:101899803-101906359 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tomt | NM_001081679.1 | 4 | 810 | NP_001075148.1 |
| NM_001282088.1 | 4 | 819 | NP_001269017.1 | |
| DQ854743.1 | 4 | 810 | ||
| EU627091.1 | 4 | 819 |
More Information
Set Membership: |
|

Genomic Map