Assay Details
Target Gene Details
Entrez Gene ID: | 654804 |
Gene Name: | RIKEN cDNA 4732471J01 gene |
Gene Aliases: |
- |
Location: |
Chr.7:25376819-25566417 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4732471J01Rik | NR_015569.3 | |||
| AK161422.1 |
Target Gene Details
Entrez Gene ID: | 26365 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 1 |
Gene Aliases: |
Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1, bb-1, mCEA1, mmCGM1, mmCGM1a, mmCGM2 |
Location: |
Chr.7:25461702-25477625 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam1 | NM_001039185.1 | 5 | 1149 | NP_001034274.1 |
| NM_001039186.1 | 5 | 1149 | NP_001034275.1 | |
| NM_001039187.1 | 3 | 609 | NP_001034276.1 | |
| NM_011926.2 | 3 | 609 | NP_036056.2 | |
| AB236328.2 | 3 | 518 | BAE45934.2 | |
| AB236329.1 | 3 | 518 | BAE45935.1 | |
| AB236330.2 | 4 | 803 | BAE45936.1 | |
| AB236331.2 | 3 | 518 | BAE45937.1 | |
| AB236332.2 | 5 | 1058 | BAE45938.1 | |
| AK085749.1 | 5 | 1109 | ||
| M77196.1 | 5 | 1127 | AAA37858.1 | |
| M96934.1 | 5 | 1075 | ||
| M96935.1 | 4 | 511 | ||
| X15351.1 | 5 | 1161 | CAA33409.1 | |
| X53084.1 | 3 | 538 | CAA37251.1 | |
| X67278.1 | 3 | 621 | CAA47695.1 | |
| X67279.1 | 5 | 1161 | CAA47696.1 | |
| X67280.1 | 5 | 1078 | CAA47697.1 | |
| X67281.1 | 5 | 1078 | CAA47698.1 | |
| X67282.1 | 3 | 621 | CAA47699.1 | |
| X67283.1 | 3 | 538 | CAA47700.1 |
More Information
Set Membership: |
|

Genomic Map