Assay Details
Target Gene Details
Entrez Gene ID: | 18301 |
Gene Name: | FXYD domain-containing ion transport regulator 5 |
Gene Aliases: |
EF-8, Oit2, RIC |
Location: |
Chr.7:31032723-31042331 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fxyd5 | NM_001111073.2 | 1 | 240 | NP_001104543.1 |
| NM_001287213.1 | NP_001274142.1 | |||
| NM_001287217.1 | NP_001274146.1 | |||
| NM_008761.4 | NP_032787.1 | |||
| AI121044.1 | ||||
| AK003068.1 | BAB22545.1 | |||
| AK155763.1 | 1 | 220 | BAE33425.1 | |
| AK169966.1 | BAE41487.1 | |||
| AK170810.1 | BAE42043.1 | |||
| CA528582.1 | ||||
| U72680.1 | AAB51040.1 |
More Information
Set Membership: |
|

Genomic Map