Assay Details
Target Gene Details
Entrez Gene ID: | 20216 |
Gene Name: | acyl-CoA synthetase medium-chain family member 3 |
Gene Aliases: |
Sa, Sah |
Location: |
Chr.7:119760883-119784896 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsm3 | NM_016870.3 | 16 | 2453 | NP_058566.3 |
| NM_212441.2 | 15 | 2348 | NP_997606.2 | |
| NM_212442.2 | 15 | 2251 | NP_997607.2 | |
| AB022340.2 | BAA37141.1 | |||
| AF068246.1 | 15 | 2238 | AAC79656.1 | |
| AK077247.1 | 15 | 2314 | ||
| AK165516.1 | 15 | 2225 | BAE38232.1 | |
| AY064696.1 | 15 | 2324 | AAL40880.1 | |
| BC015248.1 | 16 | 2413 | AAH15248.1 |
Target Gene Details
Entrez Gene ID: | 71151 |
Gene Name: | exoribonuclease 2 |
Gene Aliases: |
4933424N09Rik, Exod1, mKIAA1504 |
Location: |
Chr.7:119783826-119794058 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eri2 | NM_027698.5 | 9 | 2808 | NP_081974.3 |
| AK129376.1 | 7 | 2543 | BAC98186.1 | |
| AK135821.1 | BAE22676.1 | |||
| AK137621.1 | 9 | 2755 | ||
| AK141507.1 | 9 | 2729 | ||
| AK165163.1 | 9 | 2718 | BAE38055.1 | |
| BC090961.1 | 9 | 2710 | AAH90961.1 |
More Information
Set Membership: |
|

Genomic Map