Assay Details
Target Gene Details
Entrez Gene ID: | 330657 |
Gene Name: | protease, serine 53 |
Gene Aliases: |
Pol3s, mFLJ00289 |
Location: |
Chr.7:127885444-127890970 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss53 | NM_001081268.1 | 11 | 2388 | NP_001074737.1 |
Target Gene Details
Entrez Gene ID: | 233905 |
Gene Name: | zinc finger protein 646 |
Gene Aliases: |
6820429M01 |
Location: |
Chr.7:127877036-127885996 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp646 | NM_172749.4 | 3 | 5901 | NP_766337.2 |
| AK078520.1 | 3 | 4105 | BAC37321.1 | |
| AK129112.1 | 1 | 6277 | BAC97922.1 | |
| BC068300.1 | 2 | 5504 | AAH68300.1 | |
| BC079907.1 | 3 | 5901 | AAH79907.1 |
More Information
Set Membership: |
|

Genomic Map