Assay Details
Target Gene Details
Entrez Gene ID: | 110886 |
Gene Name: | gamma-aminobutyric acid (GABA) A receptor, subunit alpha 5 |
Gene Aliases: |
A230018I05Rik |
Location: |
Chr.7:57407669-57510100 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabra5 | NM_176942.4 | 10 | 2471 | NP_795916.1 |
| AK038476.1 | 10 | 2473 | BAC30012.1 | |
| AK083185.1 | BAC38799.1 | |||
| BC062112.1 | 10 | 2469 | AAH62112.1 |
Target Gene Details
Entrez Gene ID: | 791383 |
Gene Name: | predicted gene 9962 |
Gene Aliases: |
ENSMUSG00000055048 |
Location: |
Chr.7:57387272-57409941 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9962 | NR_033504.1 | |||
| AK049950.1 | BAC34002.1 |
More Information
Set Membership: |
|

Genomic Map