Assay Details
Target Gene Details
Entrez Gene ID: | 75736 |
Gene Name: | BCL2-like 12 (proline rich) |
Gene Aliases: |
2810475P17Rik, 5430429M05Rik, Bcl-L12, Bcl2-L12 |
Location: |
Chr.7:44991222-44997643 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l12 | NM_029410.3 | 6 | 655 | NP_083686.1 |
| AK017362.1 | 6 | 656 | BAB30709.1 |
Target Gene Details
Entrez Gene ID: | 100502630 |
Gene Name: | predicted gene 15545 |
Gene Aliases: |
OTTMUSG00000023473 |
Location: |
Chr.7:44986900-44994601 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15545 | NR_045266.1 | |||
| AK087248.1 |
More Information
Set Membership: |
|

Genomic Map