Assay Details
Target Gene Details
Entrez Gene ID: | 54399 |
Gene Name: | blocked early in transport 1 homolog (S. cerevisiae)-like |
Gene Aliases: |
2610021K23Rik, Gs15 |
Location: |
Chr.7:140853384-140856383 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bet1l | NM_018742.5 | 4 | 1340 | NP_061212.3 |
| AK003184.1 | 1 | 841 | BAB22627.1 | |
| AK011508.1 | 4 | 1225 | BAB27666.1 | |
| AK161239.1 | 4 | 1258 | BAE36260.1 |
More Information
Set Membership: |
|

Genomic Map