Assay Details
Target Gene Details
Entrez Gene ID: | 654804 |
Gene Name: | RIKEN cDNA 4732471J01 gene |
Gene Aliases: |
- |
Location: |
Chr.7:25376819-25566417 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4732471J01Rik | NR_015569.3 | |||
| AK161422.1 |
Target Gene Details
Entrez Gene ID: | 26367 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 2 |
Gene Aliases: |
AI267129, Bgp2, Ceacam2-L |
Location: |
Chr.7:25516042-25540004 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam2 | NM_001113368.1 | 2 | 367 | NP_001106839.1 |
| NM_001113369.1 | 2 | 367 | NP_001106840.1 | |
| NM_007543.4 | 2 | 367 | NP_031569.1 | |
| AB500065.1 | 2 | 192 | ||
| AB731450.1 | 2 | 127 | ||
| AF101164.1 | 2 | 225 | AAC99458.1 | |
| BC024320.1 | 2 | 176 | AAH24320.1 | |
| X76085.1 | 2 | 225 | CAA53699.1 |
More Information
Set Membership: |
|

Genomic Map