Assay Details
Target Gene Details
Entrez Gene ID: | 13446 |
Gene Name: | double C2, alpha |
Gene Aliases: |
- |
Location: |
Chr.7:126847349-126852705 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Doc2a | NM_010069.1 | 1 | 243 | NP_034199.1 |
| BC055768.1 | 1 | 243 | AAH55768.1 |

Genomic Map