Assay Details
Target Gene Details
Entrez Gene ID: | 73250 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 5 |
Gene Aliases: |
1600029H12Rik, Psg30 |
Location: |
Chr.7:17713216-17761154 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam5 | NM_028480.2 | 6 | 1814 | NP_082756.1 |
| AK145976.1 | 6 | 1815 | BAE26799.1 | |
| AK146102.1 | 6 | 1816 |
More Information
Set Membership: |
|

Genomic Map