Assay Details
Target Gene Details
Entrez Gene ID: | 12611 |
Gene Name: | CCAAT/enhancer binding protein (C/EBP), gamma |
Gene Aliases: |
C/EBP[g], C/EBPgamma, GPE1-BP, Gpe1bp, Ig/EBP |
Location: |
Chr.7:35046422-35056590 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpg | NM_009884.3 | 2 | 3209 | NP_034014.1 |
| AK087103.1 | 1 | 701 | ||
| AK171859.1 | 2 | 3116 | BAE42702.1 | |
| BC011319.1 | 2 | 3146 | AAH11319.1 |
More Information
Set Membership: |
|

Genomic Map