Assay Details
Target Gene Details
Entrez Gene ID: | 214580 |
Gene Name: | phosphoseryl-tRNA kinase |
Gene Aliases: |
5430423O14Rik, 5730458D16Rik, Gm1099, R75284 |
Location: |
Chr.7:131371113-131392768 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pstk | NM_001039534.1 | 2 | 391 | NP_001034623.1 |
| AK077585.1 | 2 | 392 | BAC36878.1 | |
| BC110381.1 | 2 | 367 | ||
| BC115503.2 | 2 | 392 |

Genomic Map