Assay Details
Target Gene Details
Entrez Gene ID: | 66356 |
Gene Name: | lysine rich nucleolar protein 1 |
Gene Aliases: |
2310008H09Rik, Tsg118 |
Location: |
Chr.7:118842217-118855998 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Knop1 | NM_001168218.1 | 2 | 692 | NP_001161690.1 |
| NM_001168219.1 | 2 | 692 | NP_001161691.1 | |
| NM_001168220.1 | 2 | 542 | NP_001161692.1 | |
| NM_023197.3 | 2 | 692 | NP_075686.2 | |
| AF034580.1 | 3 | 859 | AAD01984.1 | |
| AK009230.1 | 2 | 353 | BAB26153.1 | |
| AK083845.1 | 2 | 379 | BAC39038.1 | |
| AK166956.1 | 2 | 378 | BAE39139.1 | |
| BC055941.1 | 2 | 532 | AAH55941.1 |

Genomic Map