Assay Details
Target Gene Details
Entrez Gene ID: | 66356 |
Gene Name: | lysine rich nucleolar protein 1 |
Gene Aliases: |
2310008H09Rik, Tsg118 |
Location: |
Chr.7:118842217-118855998 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Knop1 | NM_001168218.1 | 5 | 2317 | NP_001161690.1 |
| NM_001168219.1 | 4 | 2248 | NP_001161691.1 | |
| NM_001168220.1 | 5 | 2167 | NP_001161692.1 | |
| NM_023197.3 | 5 | 2803 | NP_075686.2 | |
| AK083845.1 | 4 | 1935 | BAC39038.1 | |
| AK084607.1 | 1 | 367 |

Genomic Map