Assay Details
Target Gene Details
Entrez Gene ID: | 75099 |
Gene Name: | LysM, putative peptidoglycan-binding, domain containing 4 |
Gene Aliases: |
4930506D23Rik |
Location: |
Chr.7:67222544-67228468 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lysmd4 | NM_001191051.1 | 3 | 2862 | NP_001177980.1 |
| NM_175215.4 | 3 | 2525 | NP_780424.1 | |
| AK033681.1 | 3 | 2462 | BAC28427.1 | |
| AK172203.1 | 4 | 2625 | BAE42879.1 | |
| BC057459.1 | 3 | 2472 | AAH57459.1 |

Genomic Map