Assay Details
Target Gene Details
Entrez Gene ID: | 11859 |
Gene Name: | paired-like homeobox 2a |
Gene Aliases: |
Arix, Phox2, Pmx2, Pmx2a |
Location: |
Chr.7:101818313-101822726 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phox2a | NM_008887.2 | 1 | 332 | NP_032913.1 |
| CJ058587.1 | 1 | 333 | ||
| CJ133172.1 | 1 | 82 | ||
| X75014.1 | 1 | 174 | CAA52923.1 |
More Information
Set Membership: |
|

Genomic Map