Assay Details
Target Gene Details
Entrez Gene ID: | 69683 |
Gene Name: | ER membrane protein complex subunit 10 |
Gene Aliases: |
2310044H10Rik, 5430410O10Rik, Inm02, Mirta22 |
Location: |
Chr.7:44489938-44496524 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc10 | NM_197991.2 | 7 | 1684 | NP_932108.2 |
| AK009801.1 | ||||
| AK150499.1 | 7 | 1589 | ||
| AK160274.1 | 7 | 1601 | ||
| AK169010.1 | 7 | 1635 | BAE40807.1 | |
| AK171657.1 | BAE42592.1 | |||
| BC006065.1 | AAH06065.1 | |||
| BC020179.1 | AAH20179.1 | |||
| BY761533.1 | 1 | 467 |
More Information
Set Membership: |
|

Genomic Map