Assay Details
Target Gene Details
Entrez Gene ID: | 66824 |
Gene Name: | PYD and CARD domain containing |
Gene Aliases: |
9130417A21Rik, Asc, CARD5, TMS-1, TNS1, masc |
Location: |
Chr.7:127991373-127993867 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pycard | NM_023258.4 | 3 | 817 | NP_075747.3 |
| AB032249.1 | BAB16609.1 | |||
| AF310104.1 | AAG30287.1 | |||
| AK007742.1 | BAB25229.1 | |||
| AK009852.1 | 3 | 692 | BAB26543.1 | |
| AK018682.1 | 3 | 697 | BAB31341.1 | |
| AK152677.1 | 3 | 677 | BAE31409.1 | |
| BC008252.1 | AAH08252.1 |
More Information
Set Membership: |
|

Genomic Map