Assay Details
Target Gene Details
Entrez Gene ID: | 22793 |
Gene Name: | zyxin |
Gene Aliases: |
9530098H06Rik, R75157 |
Location: |
Chr.6:42349621-42360213 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zyx | NM_001289617.1 | 1 | 140 | NP_001276546.1 |
| NM_001289618.1 | 1 | 140 | NP_001276547.1 | |
| NM_001289619.1 | 1 | 140 | NP_001276548.1 | |
| NM_011777.3 | 1 | 140 | NP_035907.1 | |
| AK147812.1 | 1 | 141 | BAE28154.1 | |
| BC054775.1 | 1 | 99 | AAH54775.1 | |
| X99063.1 | 1 | 98 | CAA67510.1 |
More Information
Set Membership: |
|

Genomic Map