Assay Details
Target Gene Details
Entrez Gene ID: | 77582 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
5730589L02Rik, BB1, Leng4, Lpiat, Lpiat1, mBB1 |
Location: |
Chr.7:3677789-3693525 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mboat7 | NM_029934.3 | 8 | 2003 | NP_084210.2 |
| AB055410.1 | 8 | 2001 | BAC53808.1 | |
| AK019981.1 | 8 | 1414 | BAB31950.1 | |
| AK150010.1 | 8 | 1397 | BAE29235.1 | |
| AK154606.1 | 8 | 2002 | BAE32708.1 | |
| AK158998.1 | ||||
| AK170124.1 | 8 | 1480 | BAE41580.1 | |
| BC023417.1 | 8 | 1451 | AAH23417.1 | |
| BC118958.2 | 8 | 1648 | ||
| EU016380.1 | 7 | 1215 |
More Information
Set Membership: |
|

Genomic Map