Assay Details
Target Gene Details
Entrez Gene ID: | 243880 |
Gene Name: | NLR family, pyrin domain containing 4A |
Gene Aliases: |
E330021B02, E330028A19Rik, NALP4, Nalp-eta, Nalp-ita, Nalp4a |
Location: |
Chr.7:26435113-26476142 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp4a | NM_172896.2 | 11 | 3234 | NP_766484.1 |
| AK054378.1 | 11 | 3125 | ||
| AK087843.1 | 11 | 3235 | BAC40024.1 | |
| AK135745.1 | 8 | 1939 | ||
| AK162125.1 | 2 | 211 | BAE36740.1 | |
| AY355341.1 | 11 | 3235 | AAR14738.1 | |
| AY596200.1 | AAU06321.1 | |||
| BC141342.1 | 11 | 3134 | ||
| BC141343.1 | 11 | 3134 |

Genomic Map