Assay Details
Target Gene Details
Entrez Gene ID: | 210045 |
Gene Name: | NLR family, pyrin domain containing 4B |
Gene Aliases: |
AU022726, E330024M09, Nalp-gamma, Nalp4b |
Location: |
Chr.7:10687793-10730168 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp4b | NM_172481.2 | 4 | 966 | NP_766069.1 |
| AK054426.1 | 4 | 967 | BAC35775.1 | |
| AY355342.1 | 4 | 967 | AAR14739.1 | |
| AY596198.1 | 2 | 802 | AAU06319.1 | |
| BC066795.1 | 4 | 976 | AAH66795.1 |

Genomic Map