Assay Details
Target Gene Details
Entrez Gene ID: | 243996 |
Gene Name: | RIKEN cDNA 4933405O20 gene |
Gene Aliases: |
- |
Location: |
Chr.7:50599190-50600528 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933405O20Rik | NM_172901.2 | 1 | 218 | NP_766489.1 |
| AK077105.1 | 1 | 219 | BAC36614.1 | |
| BC060958.1 | 1 | 195 | AAH60958.1 |
Target Gene Details
Entrez Gene ID: | 338352 |
Gene Name: | NEL-like 1 |
Gene Aliases: |
B230343H07Rik, l7R6 |
Location: |
Chr.7:49975350-50863289 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nell1 | NM_001037906.2 | NP_001032995.1 | ||
| AY622226.1 | AAV41488.1 |
More Information
Set Membership: |
|

Genomic Map