Assay Details
Target Gene Details
Entrez Gene ID: | 16433 |
Gene Name: | CUB and zona pellucida-like domains 1 |
Gene Aliases: |
Erg-1, Itmap1, Uo-44, Usg, Utczp |
Location: |
Chr.7:131308554-131322292 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cuzd1 | NM_008411.3 | 7 | 1563 | NP_032437.3 |
| BC140992.1 | 7 | 1498 | ||
| U69699.2 | 7 | 1562 | AAC24898.2 |
More Information
Set Membership: |
|

Genomic Map