Assay Details
Target Gene Details
Entrez Gene ID: | 654804 |
Gene Name: | RIKEN cDNA 4732471J01 gene |
Gene Aliases: |
- |
Location: |
Chr.7:25376819-25566417 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4732471J01Rik | NR_015569.3 | |||
| NR_073027.1 | ||||
| AK076372.1 | ||||
| AK161422.1 |
Target Gene Details
Entrez Gene ID: | 232983 |
Gene Name: | chemokine (C-X-C motif) ligand 17 |
Gene Aliases: |
VCC-1, Vcc1 |
Location: |
Chr.7:25400053-25412886 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxcl17 | NM_153576.2 | 4 | 632 | NP_705804.2 |
| AK143005.1 | 4 | 548 | ||
| AY598463.1 | 4 | 633 | AAU04874.1 | |
| BC024561.1 | 4 | 633 | AAH24561.1 |
More Information
Set Membership: |
|

Genomic Map