Assay Details
Target Gene Details
Entrez Gene ID: | 100043254 |
Gene Name: | neuropeptide S |
Gene Aliases: |
ENSMUSG00000073804 |
Location: |
Chr.7:135268619-135272942 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nps | NM_001163611.1 | 3 | 476 | NP_001157083.1 |

Genomic Map