Assay Details
Target Gene Details
Entrez Gene ID: | 243923 |
Gene Name: | regulator of G-protein signalling 9 binding protein |
Gene Aliases: |
R9AP, Rgs9-1bp |
Location: |
Chr.7:35578993-35585582 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs9bp | NM_145840.3 | 1 | 3293 | NP_665839.1 |
| AK044285.1 | 1 | 3294 | ||
| AY229888.1 | 1 | 3255 | AAP55847.1 |

Genomic Map