Assay Details
Target Gene Details
Entrez Gene ID: | 27050 |
Gene Name: | ribosomal protein S3 |
Gene Aliases: |
D7Ertd795e, Rs_3 |
Location: |
Chr.7:99477896-99483709 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps3 | NM_012052.2 | 7 | 1702 | NP_036182.1 |
| AK164077.1 | 7 | 1645 | BAE37615.1 |
More Information
Set Membership: |
|

Genomic Map