Assay Details
Target Gene Details
Entrez Gene ID: | 70333 |
Gene Name: | CD3E antigen, epsilon polypeptide associated protein |
Gene Aliases: |
1190028F09, 2610103M17Rik, ASE-1, Ase1, CAST, PAF49 |
Location: |
Chr.7:19356007-19359483 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd3eap | NM_145822.2 | 3 | 632 | NP_665821.1 |
| AB091121.1 | 3 | 569 | BAD02839.1 | |
| AK011811.1 | 3 | 605 | ||
| BC016115.1 | 3 | 597 | ||
| BC029206.1 | 3 | 580 | AAH29206.1 | |
| BC071199.1 | 3 | 583 | AAH71199.1 |
More Information
Set Membership: |
|

Genomic Map