Assay Details
Target Gene Details
Entrez Gene ID: | 26366 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 10 |
Gene Aliases: |
Bgp3, Cea10 |
Location: |
Chr.7:24777156-24784656 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam10 | NM_007675.4 | 2 | 314 | NP_031701.3 |
| AK018529.1 | 2 | 294 | BAB31256.1 | |
| BC003346.1 | 2 | 335 | AAH03346.1 | |
| L38422.1 | 2 | 278 | AAB64011.1 |

Genomic Map