Assay Details
Target Gene Details
Entrez Gene ID: | 26366 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 10 |
Gene Aliases: |
Bgp3, Cea10 |
Location: |
Chr.7:24777156-24784656 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam10 | NM_007675.4 | 3 | 674 | NP_031701.3 |
| AK018529.1 | 3 | 654 | BAB31256.1 | |
| BC003346.1 | 3 | 695 | AAH03346.1 | |
| BE949028.1 | 2 | 387 | ||
| L38422.1 | 3 | 638 | AAB64011.1 |

Genomic Map