Assay Details
Target Gene Details
Entrez Gene ID: | 67315 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 12 |
Gene Aliases: |
1600031J20Rik, Ceacam12-C1, Ceacam12-C3, Ceacam12C1, Ceacam12C3 |
Location: |
Chr.7:18065929-18077986 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam12 | NM_001162523.1 | 3 | 849 | NP_001155995.1 |
| NM_001162524.1 | 3 | 849 | NP_001155996.1 | |
| NM_026087.3 | 3 | 849 | NP_080363.2 | |
| AK005400.1 | 3 | 851 | BAB24002.1 | |
| AK005565.1 | 3 | 800 | BAB24125.1 | |
| AK131656.1 | 3 | 839 | BAE20744.1 | |
| AK145926.1 | 3 | 800 | BAE26757.1 | |
| BC099443.1 | 3 | 787 | AAH99443.1 |
More Information
Set Membership: |
|

Genomic Map