Assay Details
Target Gene Details
Entrez Gene ID: | 67084 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 14 |
Gene Aliases: |
1600021E03Rik, 1600025E09Rik, AV033953 |
Location: |
Chr.7:17812682-17815627 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam14 | NM_025957.4 | 3 | 760 | NP_080233.1 |
| AK005390.1 | 3 | 761 | BAB23996.1 | |
| AK005482.1 | 3 | 761 | BAB24069.1 | |
| AK005533.1 | 3 | 761 | ||
| BC099412.1 | 3 | 690 | AAH99412.1 |
More Information
Set Membership: |
|

Genomic Map