Assay Details
Target Gene Details
Entrez Gene ID: | 101434 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 15 |
Gene Aliases: |
AA407838, C430002N04Rik |
Location: |
Chr.7:16671331-16675705 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam15 | NM_175315.1 | 2 | 224 | NP_780524.1 |
| AK049385.1 | 2 | 139 | BAC33726.1 | |
| BC125404.1 | 2 | 224 | ||
| BC125406.1 | 2 | 224 |
More Information
Set Membership: |
|

Genomic Map